Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-2 Kit
SDP
Catalog No. 5000079972
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human cynomolgus monkey or rhesus macaque interleukin 2 (IL-2) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of IL-2 with a lower limit of detection of 0 06 pg/mL and an upper limit of quantification of 1500 pg/mL The kit includes a 96-well plate coated with IL-2 capture antibody detection antibody read buffer calibrators and controls The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79972 Supplier Meso Scale Discovery Supplier No. K156QQG1
The Meso Scale Discovery V-PLEX Plus NHP IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human cynomolgus monkey or rhesus macaque interleukin 2 (IL-2) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of IL-2 with a lower limit of detection of 0 06 pg/mL and an upper limit of quantification of 1500 pg/mL The kit includes a 96-well plate coated with IL-2 capture antibody detection antibody read buffer calibrators and controls The assay takes approximately 3 5 hours to complete