Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-2 Kit
SDP
Catalog No. 5000080827
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of natural human interleukin 2 (IL-2) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a monoclonal antibody to human IL-2 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a 7-log dynamic range from 0 06-600 pg/mL Sample incubation time is 2 hours The kit can be used to quantify IL-2 levels and study T cell activation
Catalog No. 50-000-80827 Supplier Meso Scale Discovery Supplier No. K156QQG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus NHP IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of natural human interleukin 2 (IL-2) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a monoclonal antibody to human IL-2 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a 7-log dynamic range from 0 06-600 pg/mL Sample incubation time is 2 hours The kit can be used to quantify IL-2 levels and study T cell activation