Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-5 Kit
SDP
Catalog No. 5000079065
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-5 Kit is a highly sensitive immunoassay that enables the quantitative measurement of natural human IL-5 in serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires low sample volume The V-PLEX kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and diluents valid for serum EDTA plasma heparin plasma and culture supernatants This kit allows for the accurate quantitation of human IL-5
Catalog No. 50-000-79065 Supplier Meso Scale Discovery Supplier No. K156QSG1
The Meso Scale Discovery V-PLEX Plus NHP IL-5 Kit is a highly sensitive immunoassay that enables the quantitative measurement of natural human IL-5 in serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires low sample volume The V-PLEX kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and diluents valid for serum EDTA plasma heparin plasma and culture supernatants This kit allows for the accurate quantitation of human IL-5