Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-5 Kit
SDP
Catalog No. 5000081250
Encompass
The V-PLEX Plus NHP IL-5 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of IL-5 in non-human primate (NHP) samples such as serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescence (ECL) detection method provides high sensitivity wide dynamic range and requires low sample volumes This kit allows for the accurate reproducible quantification of NHP IL-5 across a broad range of concentrations
Catalog No. 50-000-81250 Supplier Meso Scale Discovery Supplier No. K156QSG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus NHP IL-5 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of IL-5 in non-human primate (NHP) samples such as serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescence (ECL) detection method provides high sensitivity wide dynamic range and requires low sample volumes This kit allows for the accurate reproducible quantification of NHP IL-5 across a broad range of concentrations