Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit
SDP
Catalog No. 5000079944
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a broad dynamic range from 0 06-1500 pg/mL It uses an electrochemiluminescent detection method to provide high sensitivity and wide dynamic range The kit includes all necessary reagents and can process up to 94 samples in duplicate
Catalog No. 50-000-79944 Supplier Meso Scale Discovery Supplier No. K156QXG2
The Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 06 pg/mL and a broad dynamic range from 0 06-1500 pg/mL It uses an electrochemiluminescent detection method to provide high sensitivity and wide dynamic range The kit includes all necessary reagents and can process up to 94 samples in duplicate