Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit
SDP
Catalog No. 5000080805
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit is a highly sensitive immunoassay that quantitatively measures natural human IL-6 in serum plasma and culture supernatant samples The electrochemiluminescent detection method provides sub-picogram per milliliter sensitivity and a broad dynamic range from 0 06-2500 pg/mL The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls needed to generate a standard curve and validate assay performance The V-PLEX assay requires a small sample volume and provides results in under 5 hours using an MSD instrument
Catalog No. 50-000-80805 Supplier Meso Scale Discovery Supplier No. K156QXG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus NHP IL-6 Kit is a highly sensitive immunoassay that quantitatively measures natural human IL-6 in serum plasma and culture supernatant samples The electrochemiluminescent detection method provides sub-picogram per milliliter sensitivity and a broad dynamic range from 0 06-2500 pg/mL The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls needed to generate a standard curve and validate assay performance The V-PLEX assay requires a small sample volume and provides results in under 5 hours using an MSD instrument