Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-7 Kit
SDP
Catalog No. 5000078552
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-7 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 11 pg/mL and a 7-log dynamic range from 0 49 pg/mL to 10 000 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 assays in duplicate
Catalog No. 50-000-78552 Supplier Meso Scale Discovery Supplier No. K156RCG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus NHP IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody to human IL-7 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 11 pg/mL and a 7-log dynamic range from 0 49 pg/mL to 10 000 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 assays in duplicate