Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-7 Kit
SDP
Catalog No. 5000079066
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-7 Kit is a sensitive immunoassay for measuring human IL-7 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for superior sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluents for serum EDTA plasma heparin plasma and cell culture supernatants ensuring accurate IL-7 quantification with high precision
Catalog No. 50-000-79066 Supplier Meso Scale Discovery Supplier No. K156RCG2
The Meso Scale Discovery V-PLEX Plus NHP IL-7 Kit is a sensitive immunoassay for measuring human IL-7 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for superior sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluents for serum EDTA plasma heparin plasma and cell culture supernatants ensuring accurate IL-7 quantification with high precision