Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-7 Kit
SDP
Catalog No. 5000080830
Encompass
The V-PLEX Plus NHP IL-7 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The kit includes a plate coated with a monoclonal antibody to human IL-7 a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 11 pg/mL and a range from 0 49 pg/mL to 1500 pg/mL Sample incubation time is 2 hours
Catalog No. 50-000-80830 Supplier Meso Scale Discovery Supplier No. K156RCG1
The V-PLEX Plus NHP IL-7 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The kit includes a plate coated with a monoclonal antibody to human IL-7 a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 11 pg/mL and a range from 0 49 pg/mL to 1500 pg/mL Sample incubation time is 2 hours