Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-8 Kit
SDP
Catalog No. 5000078558
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is an immunoassay that allows simultaneous measurement of 30 human cytokines and chemokines It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay takes approximately 6 hours to complete and is designed for use with serum plasma culture supernatants and other biological samples The kit can help researchers analyze cytokine networks and immune responses
Catalog No. 50-000-78558 Supplier Meso Scale Discovery Supplier No. K156RAG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is an immunoassay that allows simultaneous measurement of 30 human cytokines and chemokines It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay takes approximately 6 hours to complete and is designed for use with serum plasma culture supernatants and other biological samples The kit can help researchers analyze cytokine networks and immune responses