Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-8 Kit
SDP
Catalog No. 5000079067
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-8 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of IL-8 levels
Catalog No. 50-000-79067 Supplier Meso Scale Discovery Supplier No. K156RAG1
The Meso Scale Discovery V-PLEX Plus NHP IL-8 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of IL-8 levels