Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-8 Kit
SDP
Catalog No. 5000081548
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-8 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-8 in non-human primate serum plasma and culture supernatants The assay has a detection range of 0 06-2500 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions The kit provides high sensitivity requires a small sample volume of 50 L and has a short 2 5 hour total assay time
Catalog No. 50-000-81548 Supplier Meso Scale Discovery Supplier No. K156RAG2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Plus NHP IL-8 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-8 in non-human primate serum plasma and culture supernatants The assay has a detection range of 0 06-2500 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions The kit provides high sensitivity requires a small sample volume of 50 L and has a short 2 5 hour total assay time