Learn More
Meso Scale Discovery V-PLEX Plus NHP MCP-1 Kit
SDP
Catalog No. 5000080770
Encompass
The V-PLEX Plus NHP MCP-1 Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures monocyte chemoattractant protein 1 (MCP-1) in non-human primate serum plasma and cell culture supernatants This kit utilizes electrochemiluminescence detection technology and provides accurate reproducible results with a lower limit of detection of 19 5 pg/mL The assay requires 2 5 hours to complete and includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a 7-point calibrator curve This kit allows for the precise sensitive measurement of MCP-1 levels in non-human primate samples
Catalog No. 50-000-80770 Supplier Meso Scale Discovery Supplier No. K156NNG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus NHP MCP-1 Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures monocyte chemoattractant protein 1 (MCP-1) in non-human primate serum plasma and cell culture supernatants This kit utilizes electrochemiluminescence detection technology and provides accurate reproducible results with a lower limit of detection of 19 5 pg/mL The assay requires 2 5 hours to complete and includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a 7-point calibrator curve This kit allows for the precise sensitive measurement of MCP-1 levels in non-human primate samples