Learn More
Meso Scale Discovery V-PLEX Plus NHP MCP-1 Kit
SDP
Catalog No. 5000081143
Encompass
The V-PLEX Plus NHP MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81143 Supplier Meso Scale Discovery Supplier No. K156NNG1
The V-PLEX Plus NHP MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL