Learn More
Meso Scale Discovery V-PLEX Plus NHP MCP-4 Kit
SDP
Catalog No. 5000079942
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-plex Kit measures 30 cytokines and chemokines in cell culture supernatants serum plasma and other matrices It uses electrochemiluminescence detection for high sensitivity broad dynamic range and minimal matrix interference The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators and controls Key analytes include IL-2 IL-4 IL-6 IL-10 IL-17A/F IFN-gamma TNF-alpha and VEGF enabling efficient analysis in a small sample volume
Catalog No. 50-000-79942 Supplier Meso Scale Discovery Supplier No. K156NOG2
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-plex Kit measures 30 cytokines and chemokines in cell culture supernatants serum plasma and other matrices It uses electrochemiluminescence detection for high sensitivity broad dynamic range and minimal matrix interference The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators and controls Key analytes include IL-2 IL-4 IL-6 IL-10 IL-17A/F IFN-gamma TNF-alpha and VEGF enabling efficient analysis in a small sample volume