Learn More
Meso Scale Discovery V-PLEX Plus NHP MCP-4 Kit
SDP
Catalog No. 5000079997
Encompass
The Meso Scale Discovery MCP-4 Assay Kit is a sensitive immunoassay for measuring monocyte chemoattractant protein 4 (MCP-4) in samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a quick 2 5-hour protocol The kit includes a 96-well plate with capture and detection antibodies calibrator and diluent detecting MCP-4 as low as 3 5 pg/mL It enables reliable quantification of this chemokine biomarker involved in inflammation
Catalog No. 50-000-79997 Supplier Meso Scale Discovery Supplier No. K156NOG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery MCP-4 Assay Kit is a sensitive immunoassay for measuring monocyte chemoattractant protein 4 (MCP-4) in samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a quick 2 5-hour protocol The kit includes a 96-well plate with capture and detection antibodies calibrator and diluent detecting MCP-4 as low as 3 5 pg/mL It enables reliable quantification of this chemokine biomarker involved in inflammation