Learn More
Meso Scale Discovery V-PLEX Plus NHP MDC Kit
SDP
Catalog No. 5000078557
Encompass
The Meso Scale Discovery V-PLEX Human MDC Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage-Derived Chemokine (MDC) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay has a detection range of 19 5-2500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary MULTI-ARRAY technology and provides high sensitivity wide dynamic range and low sample volume requirements The kit includes sufficient reagents for one 96-well plate
Catalog No. 50-000-78557 Supplier Meso Scale Discovery Supplier No. K156NPG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human MDC Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage-Derived Chemokine (MDC) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay has a detection range of 19 5-2500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary MULTI-ARRAY technology and provides high sensitivity wide dynamic range and low sample volume requirements The kit includes sufficient reagents for one 96-well plate