Learn More
Meso Scale Discovery V-PLEX Plus NHP MDC Kit
SDP
Catalog No. 5000080001
Encompass
The V-PLEX Plus NHP MDC Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human cytokines chemokines growth factors and immune modulators in serum plasma and culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive plate seals It measures up to 10 analytes like IL-2 IL-4 IL-6 IL-10 IL-17A IFN- TNF- GM-CSF IL-1 and IL-12p70 using electrochemiluminescent detection on the MESO QuickPlex SQ 120 The kit s testing time is 3 5 hours for up to 80 tests
Catalog No. 50-000-80001 Supplier Meso Scale Discovery Supplier No. K156NPG2
The V-PLEX Plus NHP MDC Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human cytokines chemokines growth factors and immune modulators in serum plasma and culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive plate seals It measures up to 10 analytes like IL-2 IL-4 IL-6 IL-10 IL-17A IFN- TNF- GM-CSF IL-1 and IL-12p70 using electrochemiluminescent detection on the MESO QuickPlex SQ 120 The kit s testing time is 3 5 hours for up to 80 tests