Learn More
Meso Scale Discovery V-PLEX Plus NHP MIP-1a Kit
SDP
Catalog No. 5000081276
Encompass
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel an electrochemiluminescent multiplex immunoassay for measuring multiple cytokines chemokines growth factors and immunoglobulins from a single sample This sensitive and reproducible assay uses patented MULTI-ARRAY technology and comes in various analyte combinations The kits include all necessary reagents and plates for efficient cytokine profiling from serum plasma cell culture supernatants and other samples aiding in understanding immune response inflammation and cell signaling pathways
Catalog No. 50-000-81276 Supplier Meso Scale Discovery Supplier No. K156NQG2
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel an electrochemiluminescent multiplex immunoassay for measuring multiple cytokines chemokines growth factors and immunoglobulins from a single sample This sensitive and reproducible assay uses patented MULTI-ARRAY technology and comes in various analyte combinations The kits include all necessary reagents and plates for efficient cytokine profiling from serum plasma cell culture supernatants and other samples aiding in understanding immune response inflammation and cell signaling pathways