Learn More
Meso Scale Discovery V-PLEX Plus NHP MIP-1SY Kit
SDP
Catalog No. 5000081140
Encompass
The Meso Scale Discovery MIP-1 beta Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of Macrophage Inflammatory Protein 1 beta (MIP-1 beta) in biological samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide superior sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrator and assay diluent The assay has a typical lower limit of detection of 0 64 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81140 Supplier Meso Scale Discovery Supplier No. K156NRG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery MIP-1 beta Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of Macrophage Inflammatory Protein 1 beta (MIP-1 beta) in biological samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide superior sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrator and assay diluent The assay has a typical lower limit of detection of 0 64 pg/mL and an upper limit of quantification of 10 000 pg/mL