Learn More
Meso Scale Discovery V-PLEX Plus NHP MIP-1SY Kit
SDP
Catalog No. 5000081247
Encompass
The Meso Scale Discovery V-PLEX Plus NHP MIP-1 Kit is a multiplex immunoassay that quantitatively measures macaque MIP-1 in serum plasma and culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions The assay has a typical lower limit of detection of 1 5 pg/mL and an upper limit of quantitation of 10 000 pg/mL for macaque MIP-1
Catalog No. 50-000-81247 Supplier Meso Scale Discovery Supplier No. K156NRG2
The Meso Scale Discovery V-PLEX Plus NHP MIP-1 Kit is a multiplex immunoassay that quantitatively measures macaque MIP-1 in serum plasma and culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions The assay has a typical lower limit of detection of 1 5 pg/mL and an upper limit of quantitation of 10 000 pg/mL for macaque MIP-1