Learn More
Meso Scale Discovery V-PLEX Plus NHP TARC Kit
SDP
Catalog No. 5000081384
Encompass
The V-PLEX Plus NHP TARC Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures TARC (thymus and activation regulated chemokine) in non-human primate serum plasma and culture supernatants This kit utilizes electrochemiluminescence (ECL) detection technology and provides reagents for 96 tests in a convenient ready-to-use format With a detection range of 19 5-20 000 pg/mL and minimal required sample volume this kit enables the reliable quantification of TARC levels from small sample sizes The V-PLEX platform offers superior sensitivity a wide dynamic range and minimal matrix interference compared to traditional ELISA assays
Catalog No. 50-000-81384 Supplier Meso Scale Discovery Supplier No. K156NTG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus NHP TARC Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures TARC (thymus and activation regulated chemokine) in non-human primate serum plasma and culture supernatants This kit utilizes electrochemiluminescence (ECL) detection technology and provides reagents for 96 tests in a convenient ready-to-use format With a detection range of 19 5-20 000 pg/mL and minimal required sample volume this kit enables the reliable quantification of TARC levels from small sample sizes The V-PLEX platform offers superior sensitivity a wide dynamic range and minimal matrix interference compared to traditional ELISA assays