Learn More
Meso Scale Discovery V-PLEX Plus NHP TARC Kit
SDP
Catalog No. 5000079524
Encompass
The K156NTG-1 is a TARC ELISA kit from Meso Scale Diagnostics It is designed for the quantitative measurement of total IgE in human serum plasma and cell culture supernatant The kit utilizes MSD electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The assay time is just 3 5 hours and the kit includes a 96-well plate coated with anti-IgE capture antibody detection antibody calibrators and all necessary reagents This product enables sensitive IgE quantification for allergy research
Catalog No. 50-000-79524 Supplier Meso Scale Discovery Supplier No. K156NTG1
The K156NTG-1 is a TARC ELISA kit from Meso Scale Diagnostics It is designed for the quantitative measurement of total IgE in human serum plasma and cell culture supernatant The kit utilizes MSD electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The assay time is just 3 5 hours and the kit includes a 96-well plate coated with anti-IgE capture antibody detection antibody calibrators and all necessary reagents This product enables sensitive IgE quantification for allergy research