Learn More
Meso Scale Discovery V-PLEX Plus NHP TARC Kit
SDP
Catalog No. 5000081524
Encompass
The V-PLEX Plus NHP TARC Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures TARC (thymus and activation regulated chemokine) in non-human primate serum plasma and culture supernatants This kit utilizes electrochemiluminescence detection technology and provides reagents for 96 tests in a convenient all-in-one kit The assay has a detection range of 19 5-20 000 pg/mL and requires only 25 L of sample This kit allows for the accurate reproducible quantification of TARC levels from NHP samples
Catalog No. 50-000-81524 Supplier Meso Scale Discovery Supplier No. K156NTG2
The V-PLEX Plus NHP TARC Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures TARC (thymus and activation regulated chemokine) in non-human primate serum plasma and culture supernatants This kit utilizes electrochemiluminescence detection technology and provides reagents for 96 tests in a convenient all-in-one kit The assay has a detection range of 19 5-20 000 pg/mL and requires only 25 L of sample This kit allows for the accurate reproducible quantification of TARC levels from NHP samples