Learn More
Meso Scale Discovery V-PLEX Plus NHP TNF-SY Kit
SDP
Catalog No. 5000079017
Encompass
The V-PLEX Plus NHP TNF-beta Kit from Meso Scale Discovery is a highly sensitive immunoassay that quantitatively measures natural human TNF-beta in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide superior sensitivity dynamic range and reproducibility compared to ELISA The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the 3 5 hour assay The lower limit of detection is 0 04 pg/mL
Catalog No. 50-000-79017 Supplier Meso Scale Discovery Supplier No. K156RGG2
The V-PLEX Plus NHP TNF-beta Kit from Meso Scale Discovery is a highly sensitive immunoassay that quantitatively measures natural human TNF-beta in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide superior sensitivity dynamic range and reproducibility compared to ELISA The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the 3 5 hour assay The lower limit of detection is 0 04 pg/mL