Learn More
Meso Scale Discovery V-PLEX Plus NHP TNF-SY Kit
SDP
Catalog No. 5000079940
Encompass
The V-PLEX Plus NHP TNF-beta Kit from Meso Scale Discovery is a highly sensitive immunoassay that quantitatively measures natural human TNF-beta in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides accurate results in just 3 5 hours It requires as little as 12 5 L of sample has a lower limit of detection of 0 04 pg/mL and exhibits no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and vials of lyophilized calibrator
Catalog No. 50-000-79940 Supplier Meso Scale Discovery Supplier No. K156RGG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus NHP TNF-beta Kit from Meso Scale Discovery is a highly sensitive immunoassay that quantitatively measures natural human TNF-beta in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides accurate results in just 3 5 hours It requires as little as 12 5 L of sample has a lower limit of detection of 0 04 pg/mL and exhibits no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and vials of lyophilized calibrator