Learn More
Meso Scale Discovery V-PLEX Plus NHP TNF-SY Kit
SDP
Catalog No. 5000079968
Encompass
The K156RGG-1 is a high-performance thermoelectric module from Mesoscale using bismuth telluride alloys for efficient heating and cooling It operates up to 200 C providing 156 Watts of cooling at T 0 C with 15 4 Amps at 10 2 Volts Measuring 40 x 40 mm and 3 4 mm high it s ideal for precise temperature control in scientific medical and consumer applications A heat sink and fan are required for optimal performance The K156RGG-1 offers powerful compact thermal management
Catalog No. 50-000-79968 Supplier Meso Scale Discovery Supplier No. K156RGG1
The K156RGG-1 is a high-performance thermoelectric module from Mesoscale using bismuth telluride alloys for efficient heating and cooling It operates up to 200 C providing 156 Watts of cooling at T 0 C with 15 4 Amps at 10 2 Volts Measuring 40 x 40 mm and 3 4 mm high it s ideal for precise temperature control in scientific medical and consumer applications A heat sink and fan are required for optimal performance The K156RGG-1 offers powerful compact thermal management