Learn More
Meso Scale Discovery V-PLEX Plus NHP VEGF Kit
SDP
Catalog No. 5000079965
Encompass
The V-PLEX Plus NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of VEGF in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies specific for VEGF detection antibodies conjugated to an electrochemiluminescent label calibrators controls diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and exhibits minimal cross-reactivity It requires 2 5 hours to complete and allows high-throughput analysis of VEGF levels with as little as 25 L of sample
Catalog No. 50-000-79965 Supplier Meso Scale Discovery Supplier No. K156RHG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of VEGF in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies specific for VEGF detection antibodies conjugated to an electrochemiluminescent label calibrators controls diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and exhibits minimal cross-reactivity It requires 2 5 hours to complete and allows high-throughput analysis of VEGF levels with as little as 25 L of sample