Learn More
Meso Scale Discovery V-PLEX Plus NHP VEGF Kit
SDP
Catalog No. 5000081549
Encompass
The V-PLEX Plus NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay for measuring VEGF in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators controls diluent wash buffer read buffer and plate seals Sensitivity is 0 31 pg/mL with an upper quantification limit of 2500 pg/mL The assay takes about 3 5 hours and is used for biomarker research in angiogenesis and more
Catalog No. 50-000-81549 Supplier Meso Scale Discovery Supplier No. K156RHG2
Add to Cart
Add to Cart
The V-PLEX Plus NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay for measuring VEGF in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators controls diluent wash buffer read buffer and plate seals Sensitivity is 0 31 pg/mL with an upper quantification limit of 2500 pg/mL The assay takes about 3 5 hours and is used for biomarker research in angiogenesis and more