Learn More
Meso Scale Discovery V-PLEX Plus Rat IFN-y Kit
SDP
Catalog No. 5000081398
Encompass
The V-PLEX Plus Rat IFN-y Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying rat interferon gamma (IFN-y) in various samples It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay is sensitive to 0 3 pg/mL with a range of 0 3-10 000 pg/mL using MSD s SECTOR Imager for detection It offers high sensitivity wide range minimal interference and a 2 5-hour protocol
Catalog No. 50-000-81398 Supplier Meso Scale Discovery Supplier No. K153QOG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Rat IFN-y Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying rat interferon gamma (IFN-y) in various samples It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay is sensitive to 0 3 pg/mL with a range of 0 3-10 000 pg/mL using MSD s SECTOR Imager for detection It offers high sensitivity wide range minimal interference and a 2 5-hour protocol