Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-10 Kit
SDP
Catalog No. 5000082251
Encompass
The Meso Scale Discovery V-PLEX Rat IL-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 10 (IL-10) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of 24 4-20 000 pg/mL and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay takes 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-82251 Supplier Meso Scale Discovery Supplier No. K153QUG1
The Meso Scale Discovery V-PLEX Rat IL-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 10 (IL-10) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of 24 4-20 000 pg/mL and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay takes 3 5 hours to complete after a 2 hour incubation