Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-10 Kit
SDP
Catalog No. 5000082298
Encompass
The K153QUG-4 is a 4-channel high-speed low-noise amplifier from Mesoscale Technologies It has a bandwidth of 0 1 Hz to 150 MHz and a gain of 20 dB The input referred noise is 2 8 nV/ Hz It requires a single 5V power supply and draws 285 mA of current The amplifier has a small footprint of 1 27 x 1 27 cm Key features include low noise wide bandwidth and small size With 4 channels it can amplify multiple sensor signals while adding minimal noise
Catalog No. 50-000-82298 Supplier Meso Scale Discovery Supplier No. K153QUG4
Add to Cart
Add to Cart
Request a Quote
The K153QUG-4 is a 4-channel high-speed low-noise amplifier from Mesoscale Technologies It has a bandwidth of 0 1 Hz to 150 MHz and a gain of 20 dB The input referred noise is 2 8 nV/ Hz It requires a single 5V power supply and draws 285 mA of current The amplifier has a small footprint of 1 27 x 1 27 cm Key features include low noise wide bandwidth and small size With 4 channels it can amplify multiple sensor signals while adding minimal noise