Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-13 Kit
SDP
Catalog No. 5000082294
Encompass
The Meso Scale Discovery V-PLEX Rat IL-13 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 13 (IL-13) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for analysis on MSD instruments The assay can quantify rat IL-13 concentrations between 1 5 pg/mL - 5000 pg/mL
Catalog No. 50-000-82294 Supplier Meso Scale Discovery Supplier No. K153ODG2
The Meso Scale Discovery V-PLEX Rat IL-13 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 13 (IL-13) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for analysis on MSD instruments The assay can quantify rat IL-13 concentrations between 1 5 pg/mL - 5000 pg/mL