Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-13 Kit
SDP
Catalog No. 5000082431
Encompass
The Meso Scale Discovery V-PLEX Rat IL-13 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-13 in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for analysis on MSD instruments The assay takes 3 5 hours total time and requires a low sample volume of 50 L
Catalog No. 50-000-82431 Supplier Meso Scale Discovery Supplier No. K153ODG1
The Meso Scale Discovery V-PLEX Rat IL-13 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-13 in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for analysis on MSD instruments The assay takes 3 5 hours total time and requires a low sample volume of 50 L