Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-1SY Kit
SDP
Catalog No. 5000078338
Encompass
The Meso Scale Discovery V-PLEX Rat IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 1 beta (IL-1 ) in rat serum plasma and cell culture supernatants The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent read buffer wash buffer and plate seals The assay takes 3 5 hours to complete has a lower limit of detection of 0 04 pg/mL and a broad dynamic range from 0 04-1000 pg/mL
Catalog No. 50-000-78338 Supplier Meso Scale Discovery Supplier No. K153QPG1
The Meso Scale Discovery V-PLEX Rat IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 1 beta (IL-1 ) in rat serum plasma and cell culture supernatants The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent read buffer wash buffer and plate seals The assay takes 3 5 hours to complete has a lower limit of detection of 0 04 pg/mL and a broad dynamic range from 0 04-1000 pg/mL