Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-1SY Kit
SDP
Catalog No. 5000082427
Encompass
The Meso Scale Discovery V-PLEX Rat IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 1 beta (IL-1 ) in rat serum plasma and cell culture supernatants The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay takes approximately 3 5 hours to complete The kit can accurately quantify rat IL-1 over a range of 1 5-1000 pg/mL
Catalog No. 50-000-82427 Supplier Meso Scale Discovery Supplier No. K153QPG2
The Meso Scale Discovery V-PLEX Rat IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 1 beta (IL-1 ) in rat serum plasma and cell culture supernatants The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay takes approximately 3 5 hours to complete The kit can accurately quantify rat IL-1 over a range of 1 5-1000 pg/mL