Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-4 Kit
SDP
Catalog No. 5000082252
Encompass
The Meso Scale Discovery V-PLEX Plus Rat IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with a rat IL-4 capture antibody a ruthenium-labeled rat IL-4 detection antibody calibrators and buffers for sample dilution The assay has a detection range of 1 53-2500 pg/mL and exhibits no cross-reactivity with other cytokines The kit takes approximately 3 hours to run and only requires a small sample volume of 50 L
Catalog No. 50-000-82252 Supplier Meso Scale Discovery Supplier No. K153QRG1
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Plus Rat IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with a rat IL-4 capture antibody a ruthenium-labeled rat IL-4 detection antibody calibrators and buffers for sample dilution The assay has a detection range of 1 53-2500 pg/mL and exhibits no cross-reactivity with other cytokines The kit takes approximately 3 hours to run and only requires a small sample volume of 50 L