Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-5 Kit
SDP
Catalog No. 5000078342
Encompass
The V-PLEX Plus Rat IL-5 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The kit includes a V-PLEX Rat IL-5 Plate V-PLEX Diluent 5 V-PLEX Detection Antibody Read Buffer T (4X) and V-PLEX Sheath Fluid It has a detection range of 1 53 2500 pg/mL and requires 50 L of sample The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78342 Supplier Meso Scale Discovery Supplier No. K153QSG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Rat IL-5 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The kit includes a V-PLEX Rat IL-5 Plate V-PLEX Diluent 5 V-PLEX Detection Antibody Read Buffer T (4X) and V-PLEX Sheath Fluid It has a detection range of 1 53 2500 pg/mL and requires 50 L of sample The assay takes approximately 3 5 hours to complete