Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-5 Kit
SDP
Catalog No. 5000079812
Encompass
The Meso Scale Discovery V-PLEX Rat IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat interleukin 5 (IL-5) in cell culture supernatants serum plasma and tissue lysates The assay has a detection range of 1 5-2500 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples The assay takes approximately 3 hours to complete and has high sensitivity wide dynamic range and requires low sample volume This kit allows for the accurate quantification of rat IL-5 levels
Catalog No. 50-000-79812 Supplier Meso Scale Discovery Supplier No. K153QSG2
The Meso Scale Discovery V-PLEX Rat IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat interleukin 5 (IL-5) in cell culture supernatants serum plasma and tissue lysates The assay has a detection range of 1 5-2500 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples The assay takes approximately 3 hours to complete and has high sensitivity wide dynamic range and requires low sample volume This kit allows for the accurate quantification of rat IL-5 levels