Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-5 Kit
SDP
Catalog No. 5000081171
Encompass
The Meso Scale Discovery V-PLEX Plus Rat IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against rat IL-5 detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions used to generate a standard curve The assay has a detection range of 1 53-2500 pg/mL and exhibits no cross-reactivity with other cytokines The kit offers high sensitivity and requires a low sample volume of 50 L
Catalog No. 50-000-81171 Supplier Meso Scale Discovery Supplier No. K153QSG1
The Meso Scale Discovery V-PLEX Plus Rat IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against rat IL-5 detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions used to generate a standard curve The assay has a detection range of 1 53-2500 pg/mL and exhibits no cross-reactivity with other cytokines The kit offers high sensitivity and requires a low sample volume of 50 L