Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-6 Kit
SDP
Catalog No. 5000082312
Encompass
The V-PLEX Plus Rat IL-6 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-6 in rat serum plasma and cell culture supernatants The kit includes a V-PLEX Rat IL-6 Plate V-PLEX Diluent 5 V-PLEX Detection Antibody Read Buffer T (4X) and V-PLEX Sheath Fluid It has a detection range of 1 53-2500 pg/mL and requires 50 L of sample The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-82312 Supplier Meso Scale Discovery Supplier No. K153QXG1
The V-PLEX Plus Rat IL-6 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-6 in rat serum plasma and cell culture supernatants The kit includes a V-PLEX Rat IL-6 Plate V-PLEX Diluent 5 V-PLEX Detection Antibody Read Buffer T (4X) and V-PLEX Sheath Fluid It has a detection range of 1 53-2500 pg/mL and requires 50 L of sample The assay takes approximately 3 5 hours to complete