Learn More
Meso Scale Discovery V-PLEX Plus Rat KC/GRO Kit
SDP
Catalog No. 5000080307
Encompass
The V-PLEX Plus Rat KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of rat keratinocyte chemoattractant (KC) and rat growth-related oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized KC and GRO standards to generate standard curves and quantify sample concentrations The electrochemiluminescent assay allows sensitive and accurate quantification of KC and GRO levels with as little as 25 L of sample
Catalog No. 50-000-80307 Supplier Meso Scale Discovery Supplier No. K153QTG2
The V-PLEX Plus Rat KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of rat keratinocyte chemoattractant (KC) and rat growth-related oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized KC and GRO standards to generate standard curves and quantify sample concentrations The electrochemiluminescent assay allows sensitive and accurate quantification of KC and GRO levels with as little as 25 L of sample