Learn More
Meso Scale Discovery V-PLEX Proinflammatory Panel 1 Mouse Kit, 25 plates
SDP
Supplier:  Meso Scale Discovery K15048D4
Encompass
The Proinflammatory Panel 1 (mouse) Kit provides assay-specific components for the quantitative determination of IFN-gamma, IL-1beta, IL-2, IL-4, IL-5, IL-6, KC/GRO, IL-10, IL-12p70, and TNF-alpha. This kit may be used for the analysis of all or any combination of these cytokine biomarkers in cell culture supernatants, serum, plasma, and urine.
Catalog No. NC1269641
Only null left
Add to Cart
Request a Quote