Learn More
Meso Scale Discovery V-PLEX Proinflammatory Panel 1 Human Kit, 25 plates
SDP
Catalog No. NC1269643
Encompass
The Proinflammatory Panel 1 (human) Kit provides assay-specific components for the quantitative determination of IFN-gamma, IL-1beta, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12p70, IL-13, and TNF-alpha. This kit may be used for the analysis of all or any combination of these cytokine biomarkers in cell culture supernatants, serum, plasma, and urine.
This item is not returnable. View return policy
Catalog No. NC1269643 Supplier Meso Scale Discovery Supplier No. K15049D4
Add to Cart
Add to Cart
Request a Quote
The Proinflammatory Panel 1 (human) Kit provides assay-specific components for the quantitative determination of IFN-gamma, IL-1beta, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12p70, IL-13, and TNF-alpha. This kit may be used for the analysis of all or any combination of these cytokine biomarkers in cell culture supernatants, serum, plasma, and urine.