Learn More
Meso Scale Discovery V-PLEX Rat IFN-y Kit
SDP
Catalog No. 5000079814
Encompass
The V-PLEX Rat IFN-y Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of interferon gamma (IFN-y) in rat serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence for high sensitivity and a wide dynamic range The kit includes a 96-well plate coated with capture antibodies against rat IFN-y detection antibodies labeled with an MSD SULFO-TAG and calibrators and controls to generate a standard curve Sample incubation time is 2 hours The kit can detect IFN-y concentrations between 0 53-1500 pg/mL with an LLOQ of 0 53 pg/mL
Catalog No. 50-000-79814 Supplier Meso Scale Discovery Supplier No. K153QOD1
The V-PLEX Rat IFN-y Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of interferon gamma (IFN-y) in rat serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence for high sensitivity and a wide dynamic range The kit includes a 96-well plate coated with capture antibodies against rat IFN-y detection antibodies labeled with an MSD SULFO-TAG and calibrators and controls to generate a standard curve Sample incubation time is 2 hours The kit can detect IFN-y concentrations between 0 53-1500 pg/mL with an LLOQ of 0 53 pg/mL