Learn More
Meso Scale Discovery V-PLEX Rat IL-1SY Kit
SDP
Catalog No. 5000082295
Encompass
The V-PLEX Rat IL-1 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-1 in rat serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-82295 Supplier Meso Scale Discovery Supplier No. K153QPD1
Add to Cart
Add to Cart
The V-PLEX Rat IL-1 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-1 in rat serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete