Learn More
Meso Scale Discovery V-PLEX Rat IL-1SY Kit
SDP
Catalog No. 5000082425
Encompass
The V-PLEX Rat IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of rat interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 36 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82425 Supplier Meso Scale Discovery Supplier No. K153QPD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Rat IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of rat interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 36 pg/mL and an upper limit of quantification of 10 000 pg/mL