Learn More
Meso Scale Discovery V-PLEX Rat IL-4 Kit
SDP
Catalog No. 5000081463
Encompass
The Meso Scale Discovery V-PLEX Rat IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 4 (IL-4) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluents to generate a standard curve The assay takes 3 5 hours total time and requires a MSD instrument such as QuickPlex SQ120 or SECTOR Imager for detection
Catalog No. 50-000-81463 Supplier Meso Scale Discovery Supplier No. K153QRD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Rat IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of interleukin 4 (IL-4) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluents to generate a standard curve The assay takes 3 5 hours total time and requires a MSD instrument such as QuickPlex SQ120 or SECTOR Imager for detection