Learn More
Meso Scale Discovery V-PLEX Rat IL-4 Kit
SDP
Catalog No. 5000081560
Encompass
The Meso Scale Discovery V-PLEX Rat IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue homogenates The kit contains a 96-well plate coated with a rat IL-4 capture antibody a ruthenium-labeled rat IL-4 detection antibody calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL Sample incubation time is 2 hours
Catalog No. 50-000-81560 Supplier Meso Scale Discovery Supplier No. K153QRD2
The Meso Scale Discovery V-PLEX Rat IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue homogenates The kit contains a 96-well plate coated with a rat IL-4 capture antibody a ruthenium-labeled rat IL-4 detection antibody calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL Sample incubation time is 2 hours