Learn More
Meso Scale Discovery V-PLEX Rat IL-4 Kit
SDP
Catalog No. 5000082430
Encompass
The V-PLEX Rat IL-4 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of rat interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The assay has high sensitivity wide dynamic range and requires low sample volume It utilizes a plate that has been pre-coated with capture antibody on independent and well-defined spots The sample is added followed by detection antibody conjugated to an ECL label Results are analyzed using an MSD instrument where ECL signal is proportional to the amount of analyte present The kit includes a plate detection antibody calibrator and assay diluent
Catalog No. 50-000-82430 Supplier Meso Scale Discovery Supplier No. K153QRD1
The V-PLEX Rat IL-4 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of rat interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The assay has high sensitivity wide dynamic range and requires low sample volume It utilizes a plate that has been pre-coated with capture antibody on independent and well-defined spots The sample is added followed by detection antibody conjugated to an ECL label Results are analyzed using an MSD instrument where ECL signal is proportional to the amount of analyte present The kit includes a plate detection antibody calibrator and assay diluent